accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177839-45-1

IUPAC Name: 9,10-dibromo-2-methylanthracene
Synonyms: 9,10-dibromo-2-methylanthracene
Molecular Weight (g/mol): 350.05
Molecular Formula: C15H10Br2
InChi Key: PNDNWMMKRSTYTC-UHFFFAOYSA-N
SMILES: CC1=CC=C2C(Br)=C3C=CC=CC3=C(Br)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results