CAS RN 177842-14-7
IUPAC Name:
1-[4-(difluoromethoxy)phenyl]methanamine
Synonyms:
1-[4-(difluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol):
173.16
Molecular Formula:
C8H9F2NO
InChi Key:
UHWPLNHRROQPMB-UHFFFAOYSA-N
SMILES:
NCC1=CC=C(OC(F)F)C=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-(difluoromethoxy)benzylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.