accessibility menu, dialog, popup

UserName

Viewing profile as user:
COCOCOCOCOCOCOCOCOCOCOCOCoCoCoCo

CAS RN 17786-31-1

IUPAC Name: dodecakis(methanidylidyneoxidanium) tetracobalt
Synonyms: tetracobalt dodecacarbonyl
Molecular Weight (g/mol): 571.85
Molecular Formula: C12Co4O12
InChi Key: YMFAWOSEDSLYSZ-UHFFFAOYSA-N
SMILES: [Co].[Co].[Co].[Co].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results