accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17788-94-2

IUPAC Name: 4-bromo-4'-(4-bromophenyl)-1,1'-biphenyl
Synonyms: 4-bromo-4'-(4-bromophenyl)-1,1'-biphenyl
Molecular Weight (g/mol): 388.1
Molecular Formula: C18H12Br2
InChi Key: VAIPJQIPFPRJKJ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)