accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOO

CAS RN 17789-14-9

IUPAC Name: 2-(3-bromophenyl)-1,3-dioxolane
Synonyms: 2-(3-bromophenyl)-1,3-dioxolane
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: VYPYKCPWNPPBBX-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1OCCO1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)