accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1779-60-8

IUPAC Name: propyl 3-oxobutanoate
Synonyms: propyl 3-oxobutanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: DHGFMVMDBNLMKT-UHFFFAOYSA-N
SMILES: CCCOC(=O)CC(C)=O
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results