accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OON(R)H3N

CAS RN 177911-87-4

IUPAC Name: [(2R)-1-[(tert-butoxy)carbonyl]pyrrolidin-2-yl]methanaminium
Synonyms: [(2R)-1-(tert-butoxycarbonyl)pyrrolidin-2-yl]methanaminium
Molecular Weight (g/mol): 201.29
Molecular Formula: C10H21N2O2
InChi Key: SOGXYCNKQQJEED-MRVPVSSYSA-O
SMILES: CC(C)(C)OC(=O)N1CCC[C@@H]1C[NH3+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results