accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177966-59-5

IUPAC Name: (2S)-3-(3,4-dichlorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-(3,4-dichlorophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 456.32
Molecular Formula: C24H19Cl2NO4
InChi Key: QNVHCYWPXIGFGN-QFIPXVFZSA-N
SMILES: OC(=O)[C@H](CC1=CC=C(Cl)C(Cl)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)