accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17797-10-3

IUPAC Name: 2-(4-fluorophenyl)propan-2-amine
Synonyms: 2-(4-fluorophenyl)propan-2-amine
Molecular Weight (g/mol): 153.2
Molecular Formula: C9H12FN
InChi Key: NYFSRXOWJXAHOO-UHFFFAOYSA-N
SMILES: CC(C)(N)C1=CC=C(F)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results