CAS RN 177985-32-9
IUPAC Name:
2-(2-chloro-4-fluorophenyl)acetate
Synonyms:
2-(2-chloro-4-fluorophenyl)acetate
Molecular Weight (g/mol):
187.57
Molecular Formula:
C8H5ClFO2
InChi Key:
CDUSPKFHAVQVRQ-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CC1=CC=C(F)C=C1Cl
1
–
1
of
1
results
1
Promotions Available
2-Chloro-4-fluorophenylacetic acid, 97%
2-Chloro-4-fluorophenylacetic acid, 97%
CAS: 177985-32-9 Molecular Formula: C8H6ClFO2 Molecular Weight (g/mol): 188.582 MDL Number: MFCD00236028 InChI Key: CDUSPKFHAVQVRQ-UHFFFAOYSA-N PubChem CID: 2725061 IUPAC Name: 2-(2-chloro-4-fluorophenyl)acetic acid SMILES: C1=CC(=C(C=C1F)Cl)CC(=O)O