accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 177991-01-4

IUPAC Name: 1-ethynyl-4-[tris(4-ethynylphenyl)methyl]benzene
Synonyms: 1-ethynyl-4-[tris(4-ethynylphenyl)methyl]benzene
Molecular Weight (g/mol): 416.52
Molecular Formula: C33H20
InChi Key: JGUCYLISNORQFW-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(C=C1)C(C1=CC=C(C=C1)C#C)(C1=CC=C(C=C1)C#C)C1=CC=C(C=C1)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results