accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1780-19-4

IUPAC Name: 2-methyl-1,2,3,4-tetrahydroquinoline
Synonyms: 2-methyl-1,2,3,4-tetrahydroquinoline
Molecular Weight (g/mol): 147.22
Molecular Formula: C10H13N
InChi Key: JZICUKPOZUKZLL-UHFFFAOYNA-N
SMILES: CC1CCC2=CC=CC=C2N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results