accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1780-40-1

ClNClClClN

CAS RN 1780-40-1

IUPAC Name: tetrachloropyrimidine
Synonyms: pyrimidine, tetrachloro-
Molecular Weight (g/mol): 217.86
Molecular Formula: C4Cl4N2
InChi Key: GVBHCMNXRKOJRH-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C(Cl)C(Cl)=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)