accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1780-72-9

IUPAC Name: 4-bromo-5-methyl-1H-pyrazol-3-amine
Synonyms: 4-bromo-5-methyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 176.02
Molecular Formula: C4H6BrN3
InChi Key: WZMBDMWFVPKYCF-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(N)=NN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results