accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 178032-63-8

IUPAC Name: (1S,2R,6S,7R)-4,4-dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0²,⁶]decan-9-one
Synonyms: (1S,2R,6S,7R)-4,4-dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0²,⁶]decan-9-one
Molecular Weight (g/mol): 183.21
Molecular Formula: C9H13NO3
InChi Key: CSGIROCFQXHTKS-WNJXEPBRSA-N
SMILES: CC1(C)O[C@H]2[C@H]3C[C@@H]([C@H]2O1)C(=O)N3
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)