accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHONNHOOH3CN

CAS RN 17804-35-2

IUPAC Name: methyl N-[1-(butylcarbamoyl)-1H-1,3-benzodiazol-2-yl]carbamate
Synonyms: benomyl benlate arbortrine BBC BNM MBC fundasol NS 02 (fungicide) benomyl (substance) fundazol
Molecular Weight (g/mol): 290.32
Molecular Formula: C14H18N4O3
InChi Key: RIOXQFHNBCKOKP-UHFFFAOYSA-N
SMILES: CCCCNC(=O)N1C(NC(=O)OC)=NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results