accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 178065-29-7

IUPAC Name: 2-[(5-fluoro-2,4-dinitrophenyl)amino]-4-methylpentanamide
Synonyms: 2-[(5-fluoro-2,4-dinitrophenyl)amino]-4-methylpentanamide
Molecular Weight (g/mol): 314.27
Molecular Formula: C12H15FN4O5
InChi Key: WCOZOJGXDVGGIK-UHFFFAOYNA-N
SMILES: CC(C)CC(NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results