accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17823-58-4

CH3OOSCH3SH3CN

CAS RN 17823-58-4

IUPAC Name: ethyl 2-cyano-3,3-bis(methylsulfanyl)prop-2-enoate
Synonyms: ethyl 2-cyano-3,3-bis(methylsulfanyl)prop-2-enoate
Molecular Weight (g/mol): 217.3
Molecular Formula: C8H11NO2S2
InChi Key: POABRARINOCORV-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C#N)=C(SC)SC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results