accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17826-04-9

IUPAC Name: 6-bromo-1H-indole-3-carbaldehyde
Synonyms: 6-bromo-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: WCCLQCBKBPTODV-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(NC=C2C=O)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)