accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOOOHO

CAS RN 1783-96-6

IUPAC Name: 2-aminobutanedioic acid
Synonyms: aspartic acid 1-amino-1,2-carboxyethane tocris-0213 delta-asparaginsaeure D-aspartatic acid D aspartic acid acid D,L-aspart D aspartate delta-(-)-aspartic acid delta-aspartic acid
Molecular Weight (g/mol): 133.1
Molecular Formula: C4H7NO4
InChi Key: CKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (3)
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (6)
  • (2)
  • (7)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)