accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 178306-52-0

IUPAC Name: 2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid
Synonyms: 2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid
Molecular Weight (g/mol): 272.3
Molecular Formula: C16H16O4
InChi Key: RQJWOLFMWKZKCJ-UHFFFAOYNA-N
SMILES: COC(C(O)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results