accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOOOOOOCH2

CAS RN 17831-71-9

IUPAC Name: 2-(2-{2-[2-(prop-2-enoyloxy)ethoxy]ethoxy}ethoxy)ethyl prop-2-enoate
Synonyms: 2-(2-{2-[2-(prop-2-enoyloxy)ethoxy]ethoxy}ethoxy)ethyl prop-2-enoate
Molecular Weight (g/mol): 302.32
Molecular Formula: C14H22O7
InChi Key: HCLJOFJIQIJXHS-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCOCCOCCOCCOC(=O)C=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results