accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 178312-48-6

IUPAC Name: (4,4-difluorocyclohexyl)methanol
Synonyms: (4,4-difluorocyclohexyl)methanol
Molecular Weight (g/mol): 150.17
Molecular Formula: C7H12F2O
InChi Key: XJZNZSLOHZLFQP-UHFFFAOYSA-N
SMILES: OCC1CCC(F)(F)CC1
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)