accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOHCl

CAS RN 178312-62-4

IUPAC Name: 1,4-oxazepane hydrochloride
Synonyms: 1,4-oxazepane hydrochloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: LJRCWNIWOVZLKS-UHFFFAOYSA-N
SMILES: Cl.C1CNCCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results