accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17835-81-3

IUPAC Name: gallium(3+) triperchlorate
Synonyms: gallium(3+) triperchlorate
Molecular Weight (g/mol): 368.06
Molecular Formula: Cl3GaO12
InChi Key: DKRHELWBVMBPOQ-UHFFFAOYSA-K
SMILES: [Ga+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results