accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 178419-42-6

NHNOOHCl

CAS RN 178419-42-6

IUPAC Name: 4-azaspiro[bicyclo[2.2.2]octane-2,2'-[1,4]oxazolidin]-5'-one hydrochloride
Synonyms: 4-azaspiro[bicyclo[2.2.2]octane-2,2'-[1,4]oxazolidin]-5'-one hydrochloride
Molecular Weight (g/mol): 218.68
Molecular Formula: C9H15ClN2O2
InChi Key: XGLBLUBBDSJBIU-UHFFFAOYNA-N
SMILES: Cl.O=C1NCC2(CN3CCC2CC3)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results