accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFF

CAS RN 1785-64-4

IUPAC Name: 5,6,11,12,13,14,15,16-octafluorotricyclo[8.2.2.2⁴,⁷]hexadeca-1(12),4,6,10,13,15-hexaene
Synonyms: 5,6,11,12,13,14,15,16-octafluorotricyclo[8.2.2.2⁴,⁷]hexadeca-1(12),4,6,10,13,15-hexaene
Molecular Weight (g/mol): 352.23
Molecular Formula: C16H8F8
InChi Key: GUHKMHMGKKRFDT-UHFFFAOYSA-N
SMILES: FC1=C(F)C2=C(F)C(F)=C1CCC1=C(F)C(F)=C(CC2)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results