accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHNCH3ONHOOHOClNHOHONHOH3C

CAS RN 178603-78-6

IUPAC Name: 5-chloro-3-(2-{2-[2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}propanamido)-4-oxopentanoic acid
Synonyms: 5-chloro-3-(2-{2-[2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}propanamido)-4-oxopentanoic acid
Molecular Weight (g/mol): 541
Molecular Formula: C24H33ClN4O8
InChi Key: UOUBHJRCKHLGFB-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)C(CC1=CC=C(O)C=C1)NC(C)=O)C(=O)NC(C)C(=O)NC(CC(O)=O)C(=O)CCl
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results