accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17865-11-1

IUPAC Name: [4-(trimethylsilyl)phenyl]boronic acid
Synonyms: 4-(trimethylsilyl)phenylboronic acid
Molecular Weight (g/mol): 194.11
Molecular Formula: C9H15BO2Si
InChi Key: NRPZMSUGPMYBCQ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=C(C=C1)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results