accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiHOCH3

CAS RN 17865-32-6

IUPAC Name: (cyclohexylmethyl)dimethoxysilane
Synonyms: (cyclohexylmethyl)dimethoxysilane
Molecular Weight (g/mol): 188.34
Molecular Formula: C9H20O2Si
InChi Key: XRLIOAUFFCHSIC-UHFFFAOYSA-N
SMILES: CO[SiH](CC1CCCCC1)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)