CAS RN 17873-01-7
IUPAC Name:
trimethoxy(4-methylphenyl)silane
Synonyms:
trimethoxy(4-methylphenyl)silane
Molecular Weight (g/mol):
212.32
Molecular Formula:
C10H16O3Si
InChi Key:
XQEGZYAXBCFSBS-UHFFFAOYSA-N
SMILES:
CO[Si](OC)(OC)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
Trimethoxy(p-tolyl)silane 97.0+%, TCI America™
CAS: 17873-01-7 Molecular Formula: C10H16O3Si Molecular Weight (g/mol): 212.32 MDL Number: MFCD00048020 InChI Key: XQEGZYAXBCFSBS-UHFFFAOYSA-N PubChem CID: 2760672 IUPAC Name: trimethoxy-(4-methylphenyl)silane SMILES: CC1=CC=C(C=C1)[Si](OC)(OC)OC