accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3BrBr

CAS RN 17878-23-8

IUPAC Name: (3,5-dibromophenyl)trimethylsilane
Synonyms: (3,5-dibromophenyl)trimethylsilane
Molecular Weight (g/mol): 308.09
Molecular Formula: C9H12Br2Si
InChi Key: SZQQTWLBYAKIOT-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results