accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3Br

CAS RN 17878-47-6

IUPAC Name: (3-bromophenyl)trimethylsilane
Synonyms: (3-bromophenyl)trimethylsilane
Molecular Weight (g/mol): 229.19
Molecular Formula: C9H13BrSi
InChi Key: JXQZFRPARQBDKV-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results