accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHSOONH2

CAS RN 1788-08-5

IUPAC Name: 4-ethynylbenzene-1-sulfonamide
Synonyms: 4-ethynylbenzenesulfonamide
Molecular Weight (g/mol): 181.21
Molecular Formula: C8H7NO2S
InChi Key: OQPUCENNUFNCQO-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(C=C1)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results