accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1788-10-9

H3COSClOO

CAS RN 1788-10-9

IUPAC Name: 4-acetylbenzene-1-sulfonyl chloride
Synonyms: 4-acetylbenzenesulfonyl chloride
Molecular Weight (g/mol): 218.65
Molecular Formula: C8H7ClO3S
InChi Key: FXVDNCRTKXMSEZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)