accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1788-31-4

IUPAC Name: N-(1,3-diphenylpropan-2-ylidene)hydroxylamine
Synonyms: N-(1,3-diphenylpropan-2-ylidene)hydroxylamine
Molecular Weight (g/mol): 225.29
Molecular Formula: C15H15NO
InChi Key: SXEBLVKLMOIGER-UHFFFAOYSA-N
SMILES: ON=C(CC1=CC=CC=C1)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results