accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 178931-63-0

IUPAC Name: trimethyl({5-[5'-(trimethylstannyl)-[2,2'-bithiophen]-5-yl]thiophen-2-yl})stannane
Synonyms: trimethyl({5-[5'-(trimethylstannyl)-[2,2'-bithiophen]-5-yl]thiophen-2-yl})stannane
Molecular Weight (g/mol): 573.99
Molecular Formula: C18H24S3Sn2
InChi Key: FIKQPMKITUNTEK-UHFFFAOYSA-N
SMILES: C[Sn](C)(C)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)[Sn](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results