accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNO

CAS RN 179055-27-7

IUPAC Name: 4-(1-methyl-1H-pyrazol-3-yl)benzaldehyde
Synonyms: 4-(1-methylpyrazol-3-yl)benzaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: QWKUTEQPAJGRLZ-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results