accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 179056-79-2

CH3NNO

CAS RN 179056-79-2

IUPAC Name: 3-(1-methyl-1H-pyrazol-3-yl)benzaldehyde
Synonyms: 3-(1-methylpyrazol-3-yl)benzaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: SSAPUQDWKCUIAR-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC(C=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results