accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 179056-82-7

IUPAC Name: 4-(5-methyl-1,3,4-oxadiazol-2-yl)benzaldehyde
Synonyms: 4-(5-methyl-1,3,4-oxadiazol-2-yl)benzaldehyde
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: MPECIJQJPPLBGV-UHFFFAOYSA-N
SMILES: CC1=NN=C(O1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results