accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(R)NHCH3OOHO

CAS RN 17916-88-0

IUPAC Name: (2R)-2-acetamido-3-methylbutanoic acid
Synonyms: D-valine, N-acetyl-
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: IHYJTAOFMMMOPX-ZCFIWIBFSA-N
SMILES: CC(C)[C@@H](NC(C)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results