accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 17931-55-4

IUPAC Name: bicyclo[3.3.1]nonan-9-one
Synonyms: bicyclo[3.3.1]nonan-9-one
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: SKTMMSQPXGWCAP-UHFFFAOYSA-N
SMILES: O=C1C2CCCC1CCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results