accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SCH3H3C

CAS RN 1795-05-7

IUPAC Name: 2,3,5-trimethylthiophene
Synonyms: 2,3,5-trimethylthiophene
Molecular Weight (g/mol): 126.22
Molecular Formula: C7H10S
InChi Key: QKZJQIHBRCFDGQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results