accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1795-09-1

IUPAC Name: 2-methylthiolane
Synonyms: 2-methylthiolane
Molecular Weight (g/mol): 102.2
Molecular Formula: C5H10S
InChi Key: AJPGNQYBSTXCJE-UHFFFAOYNA-N
SMILES: CC1CCCS1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)