accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 179688-01-8

IUPAC Name: 7-(benzyloxy)-6-methoxy-1,4-dihydroquinazolin-4-one
Synonyms: 7-(benzyloxy)-6-methoxy-1H-quinazolin-4-one
Molecular Weight (g/mol): 282.3
Molecular Formula: C16H14N2O3
InChi Key: ZCUFFSHMOAEEIL-UHFFFAOYSA-N
SMILES: COC1=C(OCC2=CC=CC=C2)C=C2NC=NC(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)