accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OFFFFFFFOHCH3H3CCH3OFFFFFFFOHCH3H3CCH3OFFFFFFFOHEr(E/Z)(E/Z)(E/Z)

CAS RN 17978-75-5

IUPAC Name: tris(1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one) erbium
Synonyms: tris(1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one) erbium
Molecular Weight (g/mol): 1055.81
Molecular Formula: C30H33ErF21O6
InChi Key: LEHFMLQRGJCLOZ-UHFFFAOYSA-N
SMILES: [Er].CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results