accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOI

CAS RN 1798-06-7

IUPAC Name: 2-(4-iodophenyl)acetic acid
Synonyms: P-iodophenylacetic acid
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: FJSHTWVDFAUNCO-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results