accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1798-09-0

IUPAC Name: 2-(3-methoxyphenyl)acetate
Synonyms: 2-(3-methoxyphenyl)acetate
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H9O3
InChi Key: LEGPZHPSIPPYIO-UHFFFAOYSA-M
SMILES: COC1=CC=CC(CC([O-])=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (6)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)