accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1798-85-2

IUPAC Name: 1-bromo-3-cyclopropylbenzene
Synonyms: 1-bromo-3-cyclopropylbenzene
Molecular Weight (g/mol): 197.08
Molecular Formula: C9H9Br
InChi Key: BCBNVQYWODLZSI-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results