accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrClBrF

CAS RN 179897-90-6

IUPAC Name: 1,3-dibromo-2-chloro-5-fluorobenzene
Synonyms: 1,3-dibromo-2-chloro-5-fluorobenzene
Molecular Weight (g/mol): 288.34
Molecular Formula: C6H2Br2ClF
InChi Key: PZKDJJMHRYNBOR-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(Cl)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results